Anti-IL13RA2 Mouse Polyclonal Antibody

Lieferant: Abnova
H00003598-B01P
ABNOH00003598-B01PEA 397 EUR
ABNOH00003598-B01P
Anti-IL13RA2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR