Anti-LGALS3 Antibody

Lieferant: Abnova
H00003958-AP51
ABNOH00003958-AP51EA 842 EUR
ABNOH00003958-AP51
Anti-LGALS3 Antibody
Antikörper
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LGALS3.

Type: Primary
Antigen: LGALS3
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR