Anti-LTBR Mouse Polyclonal Antibody
Lieferant: Abnova
H00004055-B02P
ABNOH00004055-B02PEA 397 EUR
ABNOH00004055-B02P
Anti-LTBR Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LTBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human