Anti-MAK Mouse Monoclonal Antibody [clone: 4E9]

Lieferant: Abnova
H00004117-M02
ABNOH00004117-M02EA 397 EUR
ABNOH00004117-M02
Anti-MAK Mouse Monoclonal Antibody [clone: 4E9]
Antikörper
Type: Primary
Antigen: MAK
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR