Anti-MAL Mouse Polyclonal Antibody
Lieferant: Abnova
H00004118-B01P
ABNOH00004118-B01PEA 397 EUR
ABNOH00004118-B01P
Anti-MAL Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human