Anti-MYOG Mouse Monoclonal Antibody [clone: 2B7]
Lieferant: Abnova
H00004656-M01
ABNOH00004656-M01EA 397 EUR
ABNOH00004656-M01
Anti-MYOG Mouse Monoclonal Antibody [clone: 2B7]
Antikörper
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human