Anti-RAG2 Rabbit Polyclonal Antibody
Lieferant: Abnova
H00005897-D01P
ABNOH00005897-D01PEA 364 EUR
ABNOH00005897-D01P
Anti-RAG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat