Anti-SPR Polyclonal Antibody Pair

Lieferant: Abnova
H00006697-AP21
ABNOH00006697-AP21EA 842 EUR
ABNOH00006697-AP21
Anti-SPR Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human SPR.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-SPR (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-SPR (20 µg);

Type:
Antigen: SPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol SPR
Antigen-Name Surface Plasmon Resonance
Environmentally Preferable
Antigen-Synonyme SPRE_HUMAN.|SDR38C1|Sepiapterin reductase 7,8 dihydrobiopterin:NADP+ oxidoreductase|member 1|Short chain dehydrogenase/reductase family 38C|SPR
Klonalität Polyklonal
Reaktivität Human
Wirt Multiple
Gen-ID 6697
Isotyp
Lagertemperatur Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR