Anti-STK4 Mouse Monoclonal Antibody [clone: 1D7-8A10]

Lieferant: Abnova
H00006789-M01
ABNOH00006789-M01EA 397 EUR
ABNOH00006789-M01
Anti-STK4 Mouse Monoclonal Antibody [clone: 1D7-8A10]
Antikörper
Type: Primary
Antigen: STK4
Clonality: Monoclonal
Clone: 1D7-8A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR