Anti-TBX3 Mouse Monoclonal Antibody [clone: 7B3]

Lieferant: Abnova
H00006926-M08
ABNOH00006926-M08EA 397 EUR
ABNOH00006926-M08
Anti-TBX3 Mouse Monoclonal Antibody [clone: 7B3]
Antikörper
Type: Primary
Antigen: TBX3
Clonality: Monoclonal
Clone: 7B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR