Anti-HNF1B Mouse Monoclonal Antibody [clone: 4D4]

Lieferant: Abnova
H00006928-M10
ABNOH00006928-M10EA 397 EUR
ABNOH00006928-M10
Anti-HNF1B Mouse Monoclonal Antibody [clone: 4D4]
Antikörper
Type: Primary
Antigen: HNF1B
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR