Anti-TXN Antibody Pair

Lieferant: Abnova
H00007295-PW1
ABNOH00007295-PW1EA 842 EUR
ABNOH00007295-PW1
Anti-TXN Antibody Pair
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-TXN (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-TXN (50 µl)

Type:
Antigen: TXN
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR