Anti-U2AF1 Mouse Monoclonal Antibody [clone: 1C8]

Lieferant: Abnova
H00007307-M01
ABNOH00007307-M01EA 397 EUR
ABNOH00007307-M01
Anti-U2AF1 Mouse Monoclonal Antibody [clone: 1C8]
Antikörper
Type: Primary
Antigen: U2AF1
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR