Anti-XPC Mouse Monoclonal Antibody [clone: 6D11]

Lieferant: Abnova
H00007508-M01
ABNOH00007508-M01EA 397 EUR
ABNOH00007508-M01
Anti-XPC Mouse Monoclonal Antibody [clone: 6D11]
Antikörper
Type: Primary
Antigen: XPC
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR