Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 4A7]

Lieferant: Abnova
H00007546-M03
ABNOH00007546-M03EA 397 EUR
ABNOH00007546-M03
Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 4A7]
Antikörper
Type: Primary
Antigen: ZIC2 (Zic Family member 2)
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR