Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 3H7]
Lieferant: Abnova
H00007546-M08
ABNOH00007546-M08EA 397 EUR
ABNOH00007546-M08
Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 3H7]
Antikörper
Type: Primary
Antigen: ZIC2 (Zic Family member 2)
Clonality: Monoclonal
Clone: 3H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human