Anti-DNALI1 Mouse Polyclonal Antibody
Lieferant: Abnova
H00007802-B01
ABNOH00007802-B01EA 397 EUR
ABNOH00007802-B01
Anti-DNALI1 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human DNALI1 protein.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: DNALI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human