Anti-ASMTL Rabbit Polyclonal Antibody
Lieferant: Abnova
H00008623-PW1
ABNOH00008623-PW1EA 842 EUR
ABNOH00008623-PW1
Anti-ASMTL Rabbit Polyclonal Antibody
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ASMTL (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-ASMTL (50 ?l)
Type:
Antigen: ASMTL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human