Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 1C9]

Lieferant: Abnova
H00008878-M03
ABNOH00008878-M03EA 397 EUR
ABNOH00008878-M03
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 1C9]
Antikörper
Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR