Anti-HAND2 Mouse Monoclonal Antibody [clone: 4E12]

Lieferant: Abnova
H00009464-M04
ABNOH00009464-M04EA 397 EUR
ABNOH00009464-M04
Anti-HAND2 Mouse Monoclonal Antibody [clone: 4E12]
Antikörper
Type: Primary
Antigen: HAND2
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR