Anti-ATG5 Mouse Monoclonal Antibody [clone: 3G11]

Lieferant: Abnova
H00009474-M06
ABNOH00009474-M06EA 397 EUR
ABNOH00009474-M06
Anti-ATG5 Mouse Monoclonal Antibody [clone: 3G11]
Antikörper
Type: Primary
Antigen: ATG5
Clonality: Monoclonal
Clone: 3G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR