Anti-CLCA2 DNA×Pab Rabbit Polyclonal Antibody

Lieferant: Abnova
H00009635-W01P
ABNOH00009635-W01PEA 384 EUR
ABNOH00009635-W01P
Anti-CLCA2 DNA×Pab Rabbit Polyclonal Antibody
Antikörper
Type:
Antigen: CLCA2 DNAxPab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR