Anti-ISG15 Rabbit Polyclonal Antibody

Lieferant: Abnova
H00009636-D01P
ABNOH00009636-D01PEA 364 EUR
ABNOH00009636-D01P
Anti-ISG15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ISG15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR