Anti-BCL2L10 Mouse Monoclonal Antibody [clone: 1B11]

Lieferant: Abnova
H00010017-M03
ABNOH00010017-M03EA 397 EUR
ABNOH00010017-M03
Anti-BCL2L10 Mouse Monoclonal Antibody [clone: 1B11]
Antikörper
Type: Primary
Antigen: BCL2L10
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR