Anti-MCRS1 Rabbit Polyclonal Antibody

Lieferant: Abnova
H00010445-D01
ABNOH00010445-D01EA 364 EUR
ABNOH00010445-D01
Anti-MCRS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR