Anti-CAP1 Rabbit Polyclonal Antibody
Lieferant: Abnova
H00010487-D01
ABNOH00010487-D01EA 364 EUR
ABNOH00010487-D01
Anti-CAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse