Anti-PDLIM5 Mouse Polyclonal Antibody

Lieferant: Abnova
H00010611-A02
ABNOH00010611-A02EA 260 EUR
ABNOH00010611-A02
Anti-PDLIM5 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDLIM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR