Anti-POLR3F Rabbit Polyclonal Antibody

Lieferant: Abnova
H00010621-D01P
ABNOH00010621-D01PEA 364 EUR
ABNOH00010621-D01P
Anti-POLR3F Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLR3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR