Anti-LBX1 Mouse Monoclonal Antibody [clone: 2G11]
Lieferant: Abnova
H00010660-M12
ABNOH00010660-M12EA 397 EUR
ABNOH00010660-M12
Anti-LBX1 Mouse Monoclonal Antibody [clone: 2G11]
Antikörper
Type: Primary
Antigen: LBX1
Clonality: Monoclonal
Clone: 2G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human