Anti-KLF1 Mouse Monoclonal Antibody [clone: 4E10]

Lieferant: Abnova
H00010661-M02
ABNOH00010661-M02EA 397 EUR
ABNOH00010661-M02
Anti-KLF1 Mouse Monoclonal Antibody [clone: 4E10]
Antikörper
Type: Primary
Antigen: KLF1
Clonality: Monoclonal
Clone: 4E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR