Anti-LILRA2 Mouse Monoclonal Antibody [clone: 3C7]

Lieferant: Abnova
H00011027-M17
ABNOH00011027-M17EA 397 EUR
ABNOH00011027-M17
Anti-LILRA2 Mouse Monoclonal Antibody [clone: 3C7]
Antikörper
Type: Primary
Antigen: LILRA2
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR