Anti-MID2 Mouse Polyclonal Antibody

Lieferant: Abnova
H00011043-B01
ABNOH00011043-B01EA 397 EUR
ABNOH00011043-B01
Anti-MID2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR