Anti-ADAM29 Mouse Monoclonal Antibody [clone: 1F6]

Lieferant: Abnova
H00011086-M01A
ABNOH00011086-M01AEA 397 EUR
ABNOH00011086-M01A
Anti-ADAM29 Mouse Monoclonal Antibody [clone: 1F6]
Antikörper
Type: Primary
Antigen: ADAM29 (ADAM metallopeptidase domain 29)
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR