Anti-U2AF2 Mouse Monoclonal Antibody [clone: 5G8]
Lieferant: Abnova
H00011338-M03
ABNOH00011338-M03EA 397 EUR
ABNOH00011338-M03
Anti-U2AF2 Mouse Monoclonal Antibody [clone: 5G8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant U2AF2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: U2AF2
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human