Anti-ATF6 Polyclonal Antibody Pair
Lieferant: Abnova
H00022926-AP21
ABNOH00022926-AP21EA 842 EUR
ABNOH00022926-AP21
Anti-ATF6 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human ATF6.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ATF6 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-ATF6 (20 µg);
Type:
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human