Anti-UHRF1BP1L Mouse Polyclonal Antibody

Lieferant: Abnova
H00023074-B01P
ABNOH00023074-B01PEA 397 EUR
ABNOH00023074-B01P
Anti-UHRF1BP1L Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UHRF1BP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR