Anti-DAPP1 Mouse Monoclonal Antibody [clone: 1E1]

Lieferant: Abnova
H00027071-M04
ABNOH00027071-M04EA 397 EUR
ABNOH00027071-M04
Anti-DAPP1 Mouse Monoclonal Antibody [clone: 1E1]
Antikörper
Type: Primary
Antigen: DAPP1
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR