Anti-UHRF1 Mouse Monoclonal Antibody [clone: 3A11]

Lieferant: Abnova
H00029128-M01
ABNOH00029128-M01EA 397 EUR
ABNOH00029128-M01
Anti-UHRF1 Mouse Monoclonal Antibody [clone: 3A11]
Antikörper
Type: Primary
Antigen: UHRF1
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR