Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 2D5]

Lieferant: Abnova
H00051399-M03
ABNOH00051399-M03EA 397 EUR
ABNOH00051399-M03
Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 2D5]
Antikörper
Type: Primary
Antigen: TRAPPC4
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR