Anti-GSKIP Mouse Monoclonal Antibody [clone: 3A6]

Lieferant: Abnova
H00051527-M10
ABNOH00051527-M10EA 397 EUR
ABNOH00051527-M10
Anti-GSKIP Mouse Monoclonal Antibody [clone: 3A6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant C14orf129.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GSKIP
Clonality: Monoclonal
Clone: 3A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR