Anti-MTP18 Rabbit Polyclonal Antibody
Lieferant: Abnova
H00051537-PW1
ABNOH00051537-PW1EA 842 EUR
ABNOH00051537-PW1
Anti-MTP18 Rabbit Polyclonal Antibody
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-MTP18 (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-MTP18 (50 ?l)
Type:
Antigen: MTP18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human