Anti-A4GALT Mouse Monoclonal Antibody [clone: 3E12]

Lieferant: Abnova
H00053947-M08A
ABNOH00053947-M08AEA 397 EUR
ABNOH00053947-M08A
Anti-A4GALT Mouse Monoclonal Antibody [clone: 3E12]
Antikörper
Type: Primary
Antigen: A4GALT
Clonality: Monoclonal
Clone: 3E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR