Anti-GKN1 Mouse Monoclonal Antibody [clone: 1G17]

Lieferant: Abnova
H00056287-M04
ABNOH00056287-M04EA 397 EUR
ABNOH00056287-M04
Anti-GKN1 Mouse Monoclonal Antibody [clone: 1G17]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant GKN1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GKN1
Clonality: Monoclonal
Clone: 1G17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR