Anti-RETN Mouse Monoclonal Antibody (Biotin)

Lieferant: Abnova
H00056729-AP41
ABNOH00056729-AP41EA 842 EUR
ABNOH00056729-AP41
Anti-RETN Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human RETN.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-RETN, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-RETN, IgG3 kappa (50 ?g);

Type:
Antigen: RETN
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR