Anti-JAM2 Mouse Monoclonal Antibody [clone: 1C2]

Lieferant: Abnova
H00058494-M02
ABNOH00058494-M02EA 397 EUR
ABNOH00058494-M02
Anti-JAM2 Mouse Monoclonal Antibody [clone: 1C2]
Antikörper
Type: Primary
Antigen: JAM2
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR