Anti-RAB17 Polyclonal Antibody Pair

Lieferant: Abnova
H00064284-AP21
ABNOH00064284-AP21EA 842 EUR
ABNOH00064284-AP21
Anti-RAB17 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human RAB17.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-RAB17 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-RAB17 (40 µl);

Type:
Antigen: RAB17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR