Anti-STK33 Antibody Pair
Lieferant: Abnova
H00065975-AP11
ABNOH00065975-AP11EA 842 EUR
ABNOH00065975-AP11
Anti-STK33 Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human STK33.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-STK33 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-STK33, IgG2b kappa (20 µg);
Type:
Antigen: STK33
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human