Anti-PHF23 Mouse Polyclonal Antibody

Lieferant: Abnova
H00079142-B01P
ABNOH00079142-B01PEA 397 EUR
ABNOH00079142-B01P
Anti-PHF23 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human PHF23 protein.

Recommended Dilutons: Western Blot (1 µg/ml).

Type: Primary
Antigen: PHF23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR