Anti-CALN1 Mouse Monoclonal Antibody [clone: 2G5]

Lieferant: Abnova
H00083698-M29
ABNOH00083698-M29EA 397 EUR
ABNOH00083698-M29
Anti-CALN1 Mouse Monoclonal Antibody [clone: 2G5]
Antikörper
Type: Primary
Antigen: CALN1
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR