Anti-SYT16 Mouse Polyclonal Antibody
Lieferant: Abnova
H00083851-B01P
ABNOH00083851-B01PEA 397 EUR
ABNOH00083851-B01P
Anti-SYT16 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SYT16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human